siRNA Details
| Gene Symbol: Nifk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139186.2 | NP_631925.2 | 6 | 678 | ||
| XM_006249656.2 | XP_006249718.1 | 7 | 809 | |||
| GenBank mRNA | BC086585.1 | - | 6 | 678 | 7308059 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nifk| Gene Symbol: Nifk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139186.2 | NP_631925.2 | 6 | 678 | ||
| XM_006249656.2 | XP_006249718.1 | 7 | 809 | |||
| GenBank mRNA | BC086585.1 | - | 6 | 678 | 7308059 | |