siRNA Details
| Gene Symbol: Odf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024126.5 | NP_077040.2 | 2 | 801 | ||
| XM_008765448.1 | XP_008763670.1 | 2 | 609 | |||
| GenBank mRNA | BC078708.1 | - | 2 | 801 | 7114007 | |
| M58677.1 | - | 1327 | ||||
| M88759.1 | - | 2 | 818 | |||
| U09021.1 | - | 2 | 483 | |||

Odf1
Genomic Map