siRNA Details
Gene Symbol: Gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191836.1 | NP_001178765.1 | 4, 5 | 776 | ||
GenBank mRNA | S80376.1 | - | 4, 5 | 1178 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191836.1 | NP_001178765.1 | 4, 5 | 776 | ||
GenBank mRNA | S80376.1 | - | 4, 5 | 1178 |