siRNA Details
| Gene Symbol: Gnal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191836.1 | NP_001178765.1 | 10 | 1199 | ||
| GenBank mRNA | S80376.1 | - | 10 | 1601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnal| Gene Symbol: Gnal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191836.1 | NP_001178765.1 | 10 | 1199 | ||
| GenBank mRNA | S80376.1 | - | 10 | 1601 | ||