siRNA Details
| Gene Symbol: Oprd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012617.1 | NP_036749.1 | 1 | 164 | ||
| GenBank mRNA | D16348.1 | - | 1 | 164 | ||
| U00475.1 | - | 1 | 107 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oprd1| Gene Symbol: Oprd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012617.1 | NP_036749.1 | 1 | 164 | ||
| GenBank mRNA | D16348.1 | - | 1 | 164 | ||
| U00475.1 | - | 1 | 107 | |||
Set Membership: |
|