siRNA Details
| Gene Symbol: Bmf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139258.1 | NP_640351.1 | 4, 5 | 705 | ||
| XM_006234723.2 | XP_006234785.1 | 4, 5 | 743 | |||
| GenBank mRNA | AF506761.1 | - | 4, 5 | 705 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmf| Gene Symbol: Bmf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139258.1 | NP_640351.1 | 4, 5 | 705 | ||
| XM_006234723.2 | XP_006234785.1 | 4, 5 | 743 | |||
| GenBank mRNA | AF506761.1 | - | 4, 5 | 705 | ||