siRNA Details
| Gene Symbol: Slc7a6os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139328.2 | NP_647544.2 | 3 | 583 | ||
| GenBank mRNA | AY098917.2 | - | 2 | 251 | ||
| DY567141.1 | - | 3 | 583 | |||
| FQ211988.1 | - | 3 | 590 | |||
| FQ220945.1 | - | 3 | 594 | |||
| FQ224990.1 | - | 2 | 315 | |||
| FQ233662.1 | - | 3 | 592 | |||
| FQ234848.1 | - | 3 | 590 | |||

Slc7a6os
Genomic Map