siRNA Details
| Gene Symbol: Slc36a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139339.1 | NP_647555.1 | 3, 4 | 438 | ||
| GenBank mRNA | AF512430.1 | - | 3, 4 | 438 | ||
| BC166414.1 | - | 3, 4 | 372 | 9027758 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc36a2| Gene Symbol: Slc36a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139339.1 | NP_647555.1 | 3, 4 | 438 | ||
| GenBank mRNA | AF512430.1 | - | 3, 4 | 438 | ||
| BC166414.1 | - | 3, 4 | 372 | 9027758 | ||