siRNA Details
| Gene Symbol: Pnpla7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144738.2 | NP_653339.2 | 5, 6 | 590 | ||
| GenBank mRNA | BC091230.1 | - | 5, 6 | 590 | 7321805 | |
| DN934725.1 | - | 5, 6 | 574 | |||
| Gene Symbol: LOC100911867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749429.3 | XP_003749477.2 | 5, 6 | 654 | ||

Pnpla7
Genomic Map