siRNA Details
| Gene Symbol: Retn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144741.1 | NP_653342.1 | 4 | 370 | ||
| GenBank mRNA | AF378366.1 | - | 4 | 370 | ||
| AJ555618.1 | - | 3 | 189 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Retn| Gene Symbol: Retn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144741.1 | NP_653342.1 | 4 | 370 | ||
| GenBank mRNA | AF378366.1 | - | 4 | 370 | ||
| AJ555618.1 | - | 3 | 189 | |||