siRNA Details
Gene Symbol: Pdgfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031524.1 | NP_113712.1 | 6 | 1015 | ||
GenBank mRNA | Z14117.1 | - | 6 | 596 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdgfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031524.1 | NP_113712.1 | 6 | 1015 | ||
GenBank mRNA | Z14117.1 | - | 6 | 596 |