siRNA Details
| Gene Symbol: Cep104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145082.2 | NP_659550.2 | 7 | 881 | ||
| GenBank mRNA | CB544954.1 | - | 4 | 340 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep104| Gene Symbol: Cep104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145082.2 | NP_659550.2 | 7 | 881 | ||
| GenBank mRNA | CB544954.1 | - | 4 | 340 | ||