siRNA Details
| Gene Symbol: Myocd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182667.2 | NP_872608.1 | 4 | 467 | ||
| XM_006246582.2 | XP_006246644.1 | 4 | 2908 | |||
| XM_006246583.2 | XP_006246645.1 | 5 | 2952 | |||
| GenBank mRNA | AF464910.1 | - | 3 | 157 | ||

Myocd
Genomic Map