siRNA Details
| Gene Symbol: Cep350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769724.1 | XP_008767946.1 | 20 | 4816 | ||
| GenBank mRNA | AF287357.1 | - | 12 | 3303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep350| Gene Symbol: Cep350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769724.1 | XP_008767946.1 | 20 | 4816 | ||
| GenBank mRNA | AF287357.1 | - | 12 | 3303 | ||