siRNA Details
| Gene Symbol: Tp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127339.1 | NP_001120811.1 | 4 | 521 | ||
| NM_001127341.1 | NP_001120813.1 | 4 | 521 | |||
| NM_001127342.1 | NP_001120814.1 | 2 | 92 | |||
| NM_001127343.1 | NP_001120815.1 | 2 | 92 | |||
| NM_001127344.1 | NP_001120816.1 | 2 | 92 | |||
| NM_019221.3 | NP_062094.1 | 4 | 521 | |||
| XM_006248500.2 | XP_006248562.1 | 4 | 602 | |||
| XM_008768791.1 | XP_008767013.1 | 5 | 451 | |||
| GenBank mRNA | AJ277446.1 | - | 3 | 323 | ||
| AJ277447.1 | - | 2 | 92 | |||
| AJ277448.1 | - | 3 | 323 | |||
| AJ277449.1 | - | 4 | 374 | |||
| AJ277450.1 | - | 2 | 92 | |||
| AJ277451.1 | - | 4 | 374 | |||
| AJ277452.1 | - | 3 | 323 | |||
| AJ277453.1 | - | 2 | 92 | |||
| Y10258.2 | - | 4 | 521 | |||

Tp63
Genomic Map