siRNA Details
| Gene Symbol: Pnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031526.1 | NP_113714.1 | 3 | 758 | ||
| GenBank mRNA | X14211.1 | - | 3 | 663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnmt| Gene Symbol: Pnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031526.1 | NP_113714.1 | 3 | 758 | ||
| GenBank mRNA | X14211.1 | - | 3 | 663 | ||