siRNA Details
| Gene Symbol: Pomc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139326.2 | NP_647542.1 | 3 | 544 | ||
| GenBank mRNA | AF510391.1 | - | 2 | 423 | ||
| BC058443.1 | - | 3 | 544 | 6919050 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pomc| Gene Symbol: Pomc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139326.2 | NP_647542.1 | 3 | 544 | ||
| GenBank mRNA | AF510391.1 | - | 2 | 423 | ||
| BC058443.1 | - | 3 | 544 | 6919050 | ||