siRNA Details
| Gene Symbol: Ppp1cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270983.1 | NP_001257912.1 | 6 | 1039 | ||
| NM_022498.2 | NP_071943.1 | 6 | 1039 | |||
| XM_008769212.1 | XP_008767434.1 | 7 | 926 | |||
| GenBank mRNA | BC078825.1 | - | 6 | 966 | 7112668 | |
| D90165.1 | - | 6 | 868 | |||
| D90166.1 | - | 6 | 982 | |||
| FM122849.1 | - | 3 | 351 | |||
| FQ228186.1 | - | 2 | 153 | |||
| FQ232892.1 | - | 2 | 153 | |||
| M58435.1 | - | 2 | 159 | |||

Ppp1cc
Genomic Map