siRNA Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145670.4 | NP_663703.3 | 11 | 1746 | ||
| XM_008762466.1 | XP_008760688.1 | 12 | 1881 | |||
| GenBank mRNA | AB029897.1 | - | 9 | 1221 | ||
| AB180269.1 | - | 10 | 1389 | |||
| AB180270.1 | - | 8 | 1155 | |||
| AB180271.1 | - | 7 | 1113 | |||
| AB180272.1 | - | 8 | 1086 | |||
| BC094306.1 | - | 8 | 1143 | |||
| BC103480.1 | - | 10 | 1644 | |||

Bcas1
Genomic Map