siRNA Details
| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145670.4 | NP_663703.3 | 1 | 15 | ||
| XM_008762466.1 | XP_008760688.1 | 1 | 15 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcas1| Gene Symbol: Bcas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145670.4 | NP_663703.3 | 1 | 15 | ||
| XM_008762466.1 | XP_008760688.1 | 1 | 15 | |||