siRNA Details
| Gene Symbol: Ppp3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017042.2 | NP_058738.1 | 13 | 1592 | ||
| GenBank mRNA | D90036.1 | - | 13 | 1592 | ||
| FQ214219.1 | - | 638 | ||||
| M31809.1 | - | 13 | 1592 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp3cb| Gene Symbol: Ppp3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017042.2 | NP_058738.1 | 13 | 1592 | ||
| GenBank mRNA | D90036.1 | - | 13 | 1592 | ||
| FQ214219.1 | - | 638 | ||||
| M31809.1 | - | 13 | 1592 | |||