siRNA Details
| Gene Symbol: Cyp4x1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145675.1 | NP_663708.1 | 12 | 1491 | ||
| GenBank mRNA | AF439343.1 | - | 12 | 1491 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4x1| Gene Symbol: Cyp4x1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145675.1 | NP_663708.1 | 12 | 1491 | ||
| GenBank mRNA | AF439343.1 | - | 12 | 1491 | ||