siRNA Details
| Gene Symbol: Slc25a25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145677.1 | NP_663710.1 | 10 | 1356 | ||
| XM_006233931.2 | XP_006233993.1 | 11 | 1513 | |||
| XM_006233932.2 | XP_006233994.1 | 10 | 1477 | |||
| XM_006233933.2 | XP_006233995.1 | 11 | 2165 | |||
| XM_006233934.2 | XP_006233996.1 | 10 | 2129 | |||
| XM_006233935.2 | XP_006233997.1 | 11 | 2729 | |||
| XM_006233937.2 | XP_006233999.1 | 11 | 1749 | |||
| GenBank mRNA | AY043169.1 | - | 10 | 1356 | ||
| FQ211394.1 | - | 10 | 1436 | |||

Slc25a25
Genomic Map