siRNA Details
| Gene Symbol: Lrrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145679.1 | NP_663712.1 | 1 | 859 | ||
| XM_006256240.1 | XP_006256302.1 | 2 | 997 | |||
| GenBank mRNA | AB086432.1 | - | 1 | 859 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc3| Gene Symbol: Lrrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145679.1 | NP_663712.1 | 1 | 859 | ||
| XM_006256240.1 | XP_006256302.1 | 2 | 997 | |||
| GenBank mRNA | AB086432.1 | - | 1 | 859 | ||