siRNA Details
| Gene Symbol: Ppy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012626.2 | NP_036758.1 | 2 | 105 | ||
| GenBank mRNA | BP484139.1 | - | 2 | 88 | ||
| M13588.1 | - | 2 | 105 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppy| Gene Symbol: Ppy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012626.2 | NP_036758.1 | 2 | 105 | ||
| GenBank mRNA | BP484139.1 | - | 2 | 88 | ||
| M13588.1 | - | 2 | 105 | |||