siRNA Details
Gene Symbol: Prps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012634.1 | NP_036766.1 | 2, 3 | 659 | ||
GenBank mRNA | M17259.1 | - | 2, 3 | 400 | ||
M29393.1 | - | 2, 3 | 400 | |||
X16555.1 | - | 2, 3 | 659 |
Gene Symbol: LOC100910245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256848.2 | XP_006256910.2 | 2, 3 | 509 |
