siRNA Details
| Gene Symbol: Prss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012635.1 | NP_036767.1 | 1 | 25 | ||
| GenBank mRNA | FQ211333.1 | - | 1 | 28 | ||
| FQ226369.1 | - | 1 | 26 | |||
| FQ227841.1 | - | 1 | 26 | |||
| FQ233648.1 | - | 1 | 26 | |||
| FQ235029.1 | - | 1 | 26 | |||
| V01273.1 | - | 1 | 25 | |||

Prss1
Genomic Map