siRNA Details
Gene Symbol: Rarb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031529.1 | NP_113717.1 | 5, 6 | 1327 | ||
GenBank mRNA | AJ002942.1 | - | 5, 6 | 985 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rarb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031529.1 | NP_113717.1 | 5, 6 | 1327 | ||
GenBank mRNA | AJ002942.1 | - | 5, 6 | 985 |