siRNA Details
| Gene Symbol: Rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017045.1 | NP_058741.1 | 21 | 2394 | ||
| GenBank mRNA | D25233.1 | - | 21 | 2155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rb1| Gene Symbol: Rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017045.1 | NP_058741.1 | 21 | 2394 | ||
| GenBank mRNA | D25233.1 | - | 21 | 2155 | ||