siRNA Details
Gene Symbol: Rbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012640.2 | NP_036772.2 | 2 | 292 | ||
XM_006243607.1 | XP_006243669.1 | 3 | 472 | |||
GenBank mRNA | CN542437.1 | - | 3 | 379 | ||
M13949.1 | - | 2 | 292 |
