siRNA Details
| Gene Symbol: Rbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012640.2 | NP_036772.2 | 3, 4 | 397 | ||
| XM_006243607.1 | XP_006243669.1 | 4, 5 | 577 | |||
| GenBank mRNA | CN542437.1 | - | 1, 2 | 274 | ||
| M13949.1 | - | 3, 4 | 397 | |||

Rbp2
Genomic Map