siRNA Details
Gene Symbol: Ren | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012642.4 | NP_036774.4 | 6 | 748 | ||
GenBank mRNA | BC078878.1 | - | 6 | 748 | 7126969 | |
J02941.1 | - | 6 | 941 | |||
S60054.1 | - | 6 | 758 | |||
X07033.1 | - | 6 | 758 |
