siRNA Details
| Gene Symbol: RT1-N3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008855.2 | NP_001008855.1 | 8 | 1411 | ||
| XM_006255909.2 | XP_006255971.1 | 8 | 2906 | |||
| GenBank mRNA | BC158567.1 | - | 8 | 1384 | 9090368 | |
| FQ228139.1 | - | 8 | 1303 | |||
| L23128.1 | - | 8 | 1415 | |||

RT1-N3
Genomic Map