siRNA Details
| Gene Symbol: Slc2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138827.1 | NP_620182.1 | 9 | 1468 | ||
| GenBank mRNA | BC061873.1 | - | 9 | 1396 | 5598829 | |
| M13979.1 | - | 9 | 1468 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a1| Gene Symbol: Slc2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138827.1 | NP_620182.1 | 9 | 1468 | ||
| GenBank mRNA | BC061873.1 | - | 9 | 1396 | 5598829 | |
| M13979.1 | - | 9 | 1468 | |||