siRNA Details
| Gene Symbol: Sst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012659.1 | NP_036791.1 | 2 | 329 | ||
| GenBank mRNA | J00788.1 | - | 2 | 160 | ||
| M25890.1 | - | 2 | 329 | |||
| V01271.1 | - | 2 | 329 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sst| Gene Symbol: Sst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012659.1 | NP_036791.1 | 2 | 329 | ||
| GenBank mRNA | J00788.1 | - | 2 | 160 | ||
| M25890.1 | - | 2 | 329 | |||
| V01271.1 | - | 2 | 329 | |||