siRNA Details
| Gene Symbol: Thy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012673.2 | NP_036805.1 | 4 | 479 | ||
| GenBank mRNA | FQ212850.1 | - | 2 | 128 | ||
| M10666.1 | - | 2 | 329 | |||
| M18002.1 | - | 2 | 329 | |||
| X03150.1 | - | 4 | 430 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thy1| Gene Symbol: Thy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012673.2 | NP_036805.1 | 4 | 479 | ||
| GenBank mRNA | FQ212850.1 | - | 2 | 128 | ||
| M10666.1 | - | 2 | 329 | |||
| M18002.1 | - | 2 | 329 | |||
| X03150.1 | - | 4 | 430 | |||