siRNA Details
| Gene Symbol: Thy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012673.2 | NP_036805.1 | 2 | 112 | ||
| GenBank mRNA | CB797376.1 | - | 2 | 112 | ||
| X03150.1 | - | 2 | 63 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thy1| Gene Symbol: Thy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012673.2 | NP_036805.1 | 2 | 112 | ||
| GenBank mRNA | CB797376.1 | - | 2 | 112 | ||
| X03150.1 | - | 2 | 63 | |||