siRNA Details
| Gene Symbol: Tnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017056.2 | NP_058752.1 | 1 | 125 | ||
| GenBank mRNA | BC078850.1 | - | 1 | 125 | 7131725 | |
| M17096.1 | - | 1 | 126 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnp1| Gene Symbol: Tnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017056.2 | NP_058752.1 | 1 | 125 | ||
| GenBank mRNA | BC078850.1 | - | 1 | 125 | 7131725 | |
| M17096.1 | - | 1 | 126 | |||