siRNA Details
| Gene Symbol: Tp53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030989.3 | NP_112251.2 | 10 | 1240 | ||
| XM_006246594.2 | XP_006246656.1 | 11 | 1669 | |||
| XM_006246595.2 | XP_006246657.1 | 10 | 2026 | |||
| GenBank mRNA | AF209191.1 | - | 7 | 921 | ||
| AY009504.1 | - | 5 | 446 | |||
| BC081788.1 | - | 10 | 1169 | 7193583 | ||
| BC098663.1 | - | 10 | 1198 | 7384467 | ||
| FQ232659.1 | - | 10 | 1228 | |||
| U90328.1 | - | 9 | 1109 | |||
| X13058.1 | - | 9 | 1132 | |||

Tp53
Genomic Map