siRNA Details
| Gene Symbol: Wnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105715.1 | NP_001099185.1 | 2 | 218 | ||
| XM_008768262.1 | XP_008766484.1 | 2 | 328 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt3| Gene Symbol: Wnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105715.1 | NP_001099185.1 | 2 | 218 | ||
| XM_008768262.1 | XP_008766484.1 | 2 | 328 | |||