siRNA Details
| Gene Symbol: Yes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033298.1 | NP_150640.1 | 5 | 724 | ||
| XM_006245675.2 | XP_006245737.1 | 5 | 737 | |||
| GenBank mRNA | AB037472.1 | - | 4 | 501 | ||
| BC079403.1 | - | 5 | 724 | 7112783 | ||

Yes1
Genomic Map