siRNA Details
| Gene Symbol: Tbxas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012687.1 | NP_036819.1 | 8 | 909 | ||
| XM_008762759.1 | XP_008760981.1 | 8 | 970 | |||
| XM_008762760.1 | XP_008760982.1 | 6 | 2367 | |||
| XM_008762761.1 | XP_008760983.1 | 8 | 960 | |||
| GenBank mRNA | D28773.1 | - | 8 | 909 | ||
| D31798.1 | - | 8 | 831 | |||

Tbxas1
Genomic Map