siRNA Details
| Gene Symbol: Dmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005246.1 | NP_001005246.1 | 11, 12 | 1476 | ||
| XM_008773252.1 | XP_008771474.1 | 15, 16 | 1873 | |||
| XM_008773253.1 | XP_008771475.1 | 15, 16 | 1873 | |||

Dmd
Genomic Map