siRNA Details
| Gene Symbol: Ceacam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012702.2 | NP_036834.2 | 5 | 1459 | ||
| GenBank mRNA | BC098921.1 | - | 5 | 1459 | 7441743 | |
| M32474.1 | - | 5 | 1444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam3| Gene Symbol: Ceacam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012702.2 | NP_036834.2 | 5 | 1459 | ||
| GenBank mRNA | BC098921.1 | - | 5 | 1459 | 7441743 | |
| M32474.1 | - | 5 | 1444 | |||