siRNA Details
Gene Symbol: Ceacam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012702.2 | NP_036834.2 | 4 | 995 | ||
GenBank mRNA | BC098921.1 | - | 4 | 995 | 7441743 | |
M32474.1 | - | 4 | 980 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ceacam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012702.2 | NP_036834.2 | 4 | 995 | ||
GenBank mRNA | BC098921.1 | - | 4 | 995 | 7441743 | |
M32474.1 | - | 4 | 980 |