siRNA Details
| Gene Symbol: Cd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012705.1 | NP_036837.1 | 6 | 1067 | ||
| XM_006237331.2 | XP_006237393.1 | 8 | 1330 | |||
| XM_006237333.2 | XP_006237395.1 | 6 | 1090 | |||
| XM_008763280.1 | XP_008761502.1 | 6 | 1047 | |||
| GenBank mRNA | M15768.1 | - | 6 | 1067 | ||

Cd4
Genomic Map