siRNA Details
| Gene Symbol: Klra22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173291.1 | NP_775413.1 | 2 | 158 | ||
| GenBank mRNA | AY649838.1 | - | 2 | 169 | ||
| U56822.1 | - | 2 | 158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klra22| Gene Symbol: Klra22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173291.1 | NP_775413.1 | 2 | 158 | ||
| GenBank mRNA | AY649838.1 | - | 2 | 169 | ||
| U56822.1 | - | 2 | 158 | |||