siRNA Details
| Gene Symbol: Gcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012707.2 | NP_036839.2 | 4 | 332 | ||
| GenBank mRNA | BP479145.1 | - | 4 | 332 | ||
| BP480555.1 | - | 4 | 307 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gcg| Gene Symbol: Gcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012707.2 | NP_036839.2 | 4 | 332 | ||
| GenBank mRNA | BP479145.1 | - | 4 | 332 | ||
| BP480555.1 | - | 4 | 307 | |||