siRNA Details
| Gene Symbol: Glul | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017073.3 | NP_058769.4 | 2 | 134 | ||
| GenBank mRNA | BC061559.1 | - | 2 | 147 | 6918514 | |
| BC072694.1 | - | 2 | 134 | 7103110 | ||
| FQ213256.1 | - | 2 | 145 | |||
| FQ216093.1 | - | 2 | 139 | |||
| FQ226911.1 | - | 2 | 157 | |||
| FQ227474.1 | - | 2 | 146 | |||
| FQ232853.1 | - | 2 | 195 | |||
| FQ232963.1 | - | 2 | 146 | |||
| FQ233770.1 | - | 2 | 146 | |||
| FQ234289.1 | - | 2 | 147 | |||
| M29579.1 | - | 2 | 145 | |||
| M91652.1 | - | 2 | 143 | |||
| X07921.1 | - | 2 | 145 | |||

Glul
Genomic Map