siRNA Details
| Gene Symbol: LOC103690381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772785.1 | XP_008771007.1 | 1 | 41 | ||
| Gene Symbol: Lta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080769.2 | NP_542947.1 | 4 | 408 | ||

LOC103690381
Genomic Map